About this item
Recombinant CloneDetect is a fluorescein-labeled, recombinant mouse IgG with high specificity and affinity for all sub-classes of human IgG. Produced by expression in CHO cells, using no raw materials of animal origina, each batch is subjected to rigorous safety assessment, with full certification.
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA10048-866
- Antigen symbol:
- IgG
- Antigen name:
- Immunoglobulin G
- Antibody type:
- Secondary
- Reactivity: