Thinwall Polyallomer Tubes, Beckman Coulter
Catalog #: CABK331372
Supplier: Beckman Coulter
Specifications
- Dimensions:14 x 89 mm
- Volume:13.2 ml
- Supplier No.:331372
Specifications
About this item
Thinwall polyallomer tubes are for use in ultracentrifuge rotors.
They have an open-top, thinwall closure type and are transparent. Made of polyallomer material.
Customers Who Bought This Also Bought