DO308 Drying Oven, Mettler-Toledo
Catalog #: 97039-532
Supplier: Mettler Toledo
Product available on GSA Advantage®
Specifications
- Temperature Range:40 to 300 °C
- Cat. no.:97039-532
- Electrical:100/230 V
- Dimensions:300×300×550 mm
- Weight:9.1 kg
- Supplier No.:51371200
Specifications
About this item
The METTLER TOLEDO DO308 drying oven is ideal for the volumetric or coulometric Karl Fischer titrators.
- Ideal for volumetric or coulometric Karl Fischer titrators
- Easy water determination
It allows easy and accurate water determination of solid or viscous samples such as substances that release water only at elevated temperatures, substances that are poorly soluble or insoluble, substances that cause problems in the titration vessel due to their consistency, or substances that cause side reactions with Karl Fischer reagent.
Customers Who Bought This Also Bought