About this item
IntelliCAL™ ISENH3181 is a digital, combination gas-sensing ammonia ion selective electrode (ISE) with a replaceable membrane module, refillable outer body, double junction reference and built-in temperature sensor
- Provides fast, stable, and accurate results
- Ultimate traceability in measurement history
- User alerted when re-calibration is needed
- Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Special features include easy-to-replace membrane modules.
The IntelliCAL ISENH3181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENH3181 is ideal for measuring ammonia concentrations in wastewater, drinking water and general water quality applications.
Temperature range:
Continuous use; 0 to 50 °C