About this item
IntelliCAL™ ISENH4181 is a digital, combination ammonium ion selective electrode (ISE) with a non-refillable, double junction reference and built-in temperature sensor
- Utilizes a unique solid-state sensor technology that eliminates costly membrane replacements
- Requires virtually no maintenance
- Fast, stable, and accurate results
- Ultimate traceability in measurement history
- Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Special features include dry storage and fast response time. No replacement membranes.
The IntelliCAL ISENH4181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENH4181 is ideal for measuring ammonium concentrations in wastewater, drinking water and general water quality applications. The ISENH4181’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
Temperature range:
Continuous use: 0 to 50 °C