Corning® Spheroid Microplates, Corning
Supplier: Corning
Certificates
About this item
PS, black opaque microplate with optically clear bottom, with lid. The Ultra-Low Attachment surface is a covalently bound hydrogel layer that is hydrophilic and neutrally charged. The surface minimizes cell attachment, protein absorption, and enzyme activation. It is ideal for culturing stem cells, as it promotes embryoid body formation from ES cells.
- Well geometry aids formation of spheroids in centre of well
- Non cytotoxic, biologically inert and non degradable surface
- Sterilized by gamma radiation
Specifications
- Catalog No:
- 76210-918
- 10037-556
- 10037-558
- 10185-094
- 10185-096
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
Specifications
Related Information
Certificates
Search by Lot Number10037-558_14219002 10037-558_34719080 10185-094_14219050 10185-094_35219017
Conformity declaration
Customers Who Bought This Also Bought