1-Nitropropane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Certificates
About this item
Formula:C₃H₇NO₂
MW:89.09 g/mol
MDL Number:MFCD00007407
CAS Number:108-03-2
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ200
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: -108 °C
:Flash Pt:
:Flash Pt:
:M.W. 89.09 g/mol
:1-Nitropropane
:-108
:131
UN:2608
:3
:III
:36
:GHS02
:GHS07
:MQ200
:12,06724
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Sigma-Aldrich®, 1-Nitropropane for synthesis:
- Solubility:14 g/l (20 °C)
- Melting Point:-108 °C
- Molar Mass:89.09 g/mol
- Boiling Point:131 - 132 °C (1013 hPa)
- Vapor Pressure:7.6 hPa (20 °C)
- Flash Point:36 °C
- Refractive Index:1.4016 (20 °C, 589 nm)
- Explosion Limit:2.2 - 11.0 %(V)
- Density:0.99 g/cm3 (20 °C)
- pH:6.0 (0.9 g/l, H2O, 20 °C)
- Ignition Point:420 °C
Specification Test Results
Related Information
Customers Who Bought This Also Bought