Sodium hydrogen carbonate/sodium carbonate certified secondary standard reference material for pH measurement; directly traceable to primary SRM from NIST/PTB pH(S)=10.014 (25°C) (DIN 19266) Certipur®
Supplier: MERCK, S.A. DE C.V. SV
Certificates
About this item
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.