ContactUsLinkComponent
Pluro Stoppers, Kimble Chase
Pluro Stoppers, Kimble Chase
Catalog #: 46600-788
PLURO STOPPER SET OF 7 = 1PK
Pluro Stoppers, Kimble Chase
Catalog #: 46600-788
Specifications
  • Description:
    Set of 7 Neoprene Rubber Pluro Stoppers
  • Cat. no.:
    46600-788
  • Supplier No.:
    852050-0070
Specifications
About this item
This autoclavable neoprene rubber adapter set consists of seven cones designed to fit buchner and fritted glass funnels
Set includes one of each sizes 1 through 7.
Can be used singly, or nested with adjacent sizes. Eliminates boring of special size holes in rubber stoppers and difficulties of insertion and removal.
Customers Who Bought This Also Bought