Amplitube™ Thin Wall PCR Reaction Strips, Simport Scientific
Supplier: Simport Scientific
Certificates
About this item
These 8-tube reaction strips feature attached 8-cap strips molded with hinge to faciliatate opening and closing.
- Made of polypropylene
- Autoclavable
Domed cap design offers a snap shut seal to avoid evaporation during thermal cycling stages.
Certified RNase-, DNase-, Pyrogen-, and DNA-free.
Packaging: Strips are packed in tamper-proof resealable bags.
Customers Who Bought This Also Bought