Amplitube™ Low-Profile PCR Reaction Strips, Simport Scientific
Supplier: Simport Scientific
Certificates
About this item
Low-profile, 100 µl strip of 8 tubes with individually attached hinged flat caps.
- Strips are manufactured from polypropylene
Certified RNase-, DNase-, Pyrogen-, and DNA-free.
Customers Who Bought This Also Bought