Amplitube™ PCR Reaction Tubes, 0.5 ml, Simport Scientific
Supplier: Simport Scientific
Certificates
About this item
Tubes are made of transparent superior grade polypropylene for improved viewing of the sample contents.
- Attached hinged caps are either dome or flat-topped
- Caps provide positive sealing during thermal cycling and prevent evaporation
- Caps can be easily opened and closed with one hand
- Graduated in 0.1 ml increments
- Autoclavable
Tubes also feature an integral shield that prevents contamination with surface of lid. A frosted writing surface is provided for sample identification.
Ultra-thin wall design ensures rapid thermal transfer and significant reduction in cycle and PCR reaction time.
Rnase-, Dnase-, Pyrogen-, and DNA-free.
Ordering information: Available in colorless or four non-cytotoxic colors.
Packaging: Packaged in tamper-proof, resealable safety-lock bags.
Customers Who Bought This Also Bought