Ultrasonic Booster, Ace Glass Incorporated
Catalog #: 89075-516
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Ultrasonic Booster
- Cat. no.:89075-516
Specifications
About this item
When connected between the converter and horn, the booster increases the amplitude of vibration at the horn tip by a factor of two
Use to process very difficult applications
Customers Who Bought This Also Bought