ContactUsLinkComponent
SP Bel-Art Conway Diffusion Cell, Bel-Art Products, a part of SP
SP Bel-Art Conway Diffusion Cell, Bel-Art Products, a part of SP
Catalog #: 70690-140
CELL, PP, CONWAY DIFFUSION, 3/PKG, 36/CASE
SP Bel-Art Conway Diffusion Cell, Bel-Art Products, a part of SP
Catalog #: 70690-140
Specifications
  • O.D.:
    83 mm
  • Description:
    Conway Diffusion Cell
  • Cat. no.:
    70690-140
  • Supplier No.:
    409410000
Specifications
About this item
Designed for determining carbon dioxide, ammonia, or alcohol in blood or serum by diffusion method.
  • White, molded polypropylene construction
  • Natural, clear polypropylene cover for extra visibility
  • Autoclavable
Smooth, white, molded polypropylene with a natural, clear polypropylene cover
The Diffusion Cell has a moat depth of 4 mm. The chamber depth is 10 mm, with a 60 mm diameter. The center depth is 4 mm with a 35 mm diameter.
The cover has a lifting knob and fits the annular moat of the cell to seal off the chamber. Steam autoclavable at 121°C (250°F).
Customers Who Bought This Also Bought