SP Bel-Art Bel-Blotter™ 96-Well Replicating Tool, Bel-Art Products, a part of SP
Catalog #: 47750-528
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Bel-Blotter™ 96-Well Replicating Tool
- Cat. no.:47750-528
- Supplier No.:378760002
Specifications
About this item
Designed to perform tedious tasks with minimum effort, the 96 open-ended pipet tips of this unique product fit all types of 96 well plates from flat, "V", or round bottom plates to 0.2 mL thin walled PCR plates and tubes.
- Suited for repetitive routines
- Polycarbonate construction
- Easy to use
- Reusable
- Autoclavable
The tip will retain up to 10 mL of fluid for placement onto filter paper, plate, or other receiving media.
The tool can be used for replication of recombinant DNA libraries, inoculation of filters for colony hybridization, PCR, phage typing, and other applications. Made of polycarbonate, it is easy-to-use, reusable and autoclavable.
Customers Who Bought This Also Bought