Stirring Shaft with PTFE Agitator, 5 mm, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Stirring shaft agitator combination available in either a solid, precision ground glass shaft or all PTFE knob type shaft with a PTFE snap-fit agitator designed to fit through standard taper 14/20 and spherical 18/11 joints
Designed for use with 5 mm I.D. bearings and flasks of 50 mL and smaller.
Customers Who Bought This Also Bought