SP Bel-Art Flowmi™ Cell Strainers for 1000 µl Pipet Tips, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Scienceware® Flowmi™ cell strainers provide fast and efficient filtering of small volume samples (up to 1000 µl) prior to FLOW or FACS analysis. Patent pending.
- Preserves volume when filtering small sample volumes
- Removes cell aggregates and creates uniform single cell suspensions
- Decreases the potential for clogging of FLOW or FACS instruments by effectively filtering cellular debris
- Sterile, 40 µm or 70 µm porosity
Flowmi™ is recommended for use with samples having a maximum concentration of 2 million cells/ml (40 µm) or 4 million cells/ml (70 µm). Use with pipette tips; Flowmi™ fits most 1000 µl tips including Fisherbrand Sure-One, VWR®, Axygen, Nichiryo and Eppendorf and more. Compact tray holds 50 Flowmi™ Cell Strainers ready for press and fit attachment to your tip; features a sliding cover for easy, one-handed use.
Packaging: Packaged in a re-sealable bag to maintain sterility when used in a laminar flow hood or biohazard hood.
Caution: Sterilized products are not returnable.