Button Type Stirring Shaft, 5 mm, Hollow Glass, Ace Glass Incorporated
Catalog #: 89075-084
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Hollow Glass Stirring Shaft
- Cat. no.:89075-084
- Shaft Diameter:5 mm
- Supplier No.:9535-06
Specifications
About this item
Precision ground, hollow, 5 mm glass stirring shaft
Button type shaft accepts blades and can be used in matching bearings. Holes are provided in the side of the shaft for material introduction and at the bottom for emission into the flask contents. Join to stirrer motor using certain Ace series chucks.
Customers Who Bought This Also Bought