Precision Ground Stirring Shaft, Ace Glass
Catalog #: 89075-076
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Borosilicate Glass Stirring Shaft, Precision Ground, 10 mm
- Cat. no.:89075-076
- Supplier No.:9533-02
Specifications
About this item
Precision ground glass stirring shaft, 10 mm O.D., attached vanes
Vanes will fit through a standard taper 45/50 joint and are contoured for use with Ace series Mini-Lab flasks. Use with Trubore® bearings. Compatible with Ace mechanical seal and PTFE collet type bearings. Join to stirrer motor using series chucks.
Customers Who Bought This Also Bought