SP Bel-Art Fritware® Immersion Filters, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
For filtration involving recovery of solids without having to transfer material to funnels.
- HDPE construction
- Available in a range of sizes
- Can be precoated for fine filtration
High-density polyethylene plastic coarse porosity disc (90-130 micron) and body. Use directly or precoat for fine filtration. Larger filter has a serrated tubing connector.
Customers Who Bought This Also Bought