Ultrasonic Reactor, Large Volume, Ace Glass Incorporated
Catalog #: 89075-526
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Complete Reaction Assembly
- Cat. no.:89075-526
- Supplier No.:9831-40
Specifications
About this item
Complete reaction assembly with parts necessary to perform ultrasonic reactions and mixing from 250 to 1800 mL
Includes three borosilicate reactors, power supply with converter, 3/4" horn, 3/4" extender, slide adapter and clamp.
Customers Who Bought This Also Bought