Circulator Hose Clamp, Ace Glass Incorporated
Catalog #: 89083-196
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Adjustable Circulator Hose Clamp
- Cat. no.:89083-196
- Supplier No.:13010-01
Specifications
About this item
All stainless steel, fully adjustable clamps for securing and supporting circulator hoses used with jacketed pilot plants
The universal clamps adjust to any pipe style pilot plant stand, and the clip will accommodate any standard circulator hose size.
Customers Who Bought This Also Bought