Direct-Drive Two-Stage Vacuum Pump, 85 LPM, Ace Glass Incorporated
Catalog #: 89078-176
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Direct-Drive Two-Stage Vacuum Pump
- Cat. no.:89078-176
- Supplier No.:14026-20
Specifications
About this item
Two-stage, direct-drive pump which provides the best performance/cost ratios for industrial or laboratory applications
Rotary sliding vane gives 85 liter per minute (3 cfm) capacity and operates quietly. Features a gas ballast to help purge moisture from the vacuum system and trap that reduces the risk of oil suck back.
Customers Who Bought This Also Bought