StrataCooler LP Benchtop Cooler, Agilent Technologies
Catalog #: CAST401349
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Specifications
- Color:Blue
- Capacity:32× 1.5 ml
- Dimensions:18.7×12×8 cm
Specifications
About this item
The StrataCooler LP benchtop cooler provides the most protection for enzymes whether in freezer or on benchtop.
- Maintains –15 °C for at least 2 hours
- Eliminates ice chips and potential contamination
Cooler with slots for holding 1.5 ml and 0.5 ml tubes to keep temperature-sensitive enzymes cold while working at the bench.
Delivery: Cooler includes chilled lid, adapters and a locking handle.
Customers Who Bought This Also Bought