Universal Valve Tubing Connector, Ace Glass Incorporated
Catalog #: 89077-466
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Teflon Connection Valve Only
- Cat. no.:89077-466
- Supplier No.:12735-15
Specifications
About this item
This universal tubing connector body includes an integral linear valve designed to provide flow control while connecting large O.D tubing to small O.D tubing.
The OMNIFIT flangeless principle applies to both large and small tubing connections. The connector body is PTFE. The large end cap is Delrin. The smaller end cap is polypropylene and available in ten colors or Tefzel with 1/4"-28. thread. Valve key is Tefzel. The unit may be cleaned with strong mineral acids and is autoclavable.
Customers Who Bought This Also Bought