Flush Seal Bottom Drain Valve Assembly with Pin, Ace Glass Incorporated
Catalog #: 71001-526
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:PTFE Valve Assembly, Flush Seal with Top Pin, Complete Valve Assembly 1" Beaded Pipe
- Cat. no.:71001-526
- Supplier No.:6482-20
Specifications
About this item
Redesigned flush-seal type valve with the top of the valve having a 1-1/2" x 3/8" O.D
pin that extends upward. The pin fits into the receptacle on the bottom of certain ACE agitators. This assembly is designed to reduce shaft wobble or “flexing” on the larger reactors. Top opening is 2-inch beaded pipe and the side drain port is 1-inch beaded pipe. Uses clamp from product ID#89064-666.
Customers Who Bought This Also Bought