Micro/Mini-Lab Vacuum Take-Off Adapter with Hose Connection, Ace Glass Incorporated
Catalog #: 89075-148
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Cat. no.:89075-148
- Bottom Inner [ST] Joint Size:14/10
- Top Outer ST Joint Size:[ST] 14/10
- Supplier No.:9564-10
Specifications
About this item
Adapter is used for microscale vacuum distillations.
- Top joint is a combination cap, thread and [ST] joint
- Bottom inner joint
- With hose connection for vacuum
- 105° angle
- Cap and O-Ring included
The top joint is a combination cap, thread and standard taper joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external thread to facilitate an O-Ring and open-top phenolic cap. The internal or I.D. at the top is a ground standard taper 14/10 outer joint. Inner joint at other end and size C hose connection for vacuum. 105° angle. Supplied with cap and O-Ring.
Customers Who Bought This Also Bought