Micro/Mini-Lab Funnel, Ace Glass Incorporated
Catalog #: 89075-472
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Cat. no.:89075-472
- Bottom Inner [ST] Joint Size:14/10
- Supplier No.:9729-21
Specifications
About this item
Comes with standard taper inner joint
Top diameter is 40 mm. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought