Micro/Mini-Lab Distilling Adapter, Ace Glass Incorporated
Catalog #: 89075-144
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Cat. no.:89075-144
- Bottom [ST] Joint Size:14/10
- Top ST Joint Size:[ST] 14/10
- Supplier No.:9562-11
Specifications
About this item
Adapter's top joint is a combination cap, thread and joint to accommodate and seal other microscale apparatus.
- Top joint is a combination cap, thread and joint
- Bottom [ST] inner joint
- 75° angle
- Cap and O-Ring included
The O.D. of the top joint has an external thread to facilitate an O-Ring and open-top phenolic cap. The internal or I.D. at the top is a ground standard taper 14/10 outer joint. A standard taper 14/10 inner joint at bottom and at 75° angle. Supplied with cap and O-Ring.
Customers Who Bought This Also Bought