Micro/Mini-Lab Cap Stopper, Ace Glass Incorporated
Catalog #: 89075-474
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- ST Joint No.:14/10
- Cat. no.:89075-474
- Supplier No.:9730-10
Specifications
About this item
With a standard taper 14/10 outer joint externally threaded for making O-Ring Cap Seal connection with mating inner joint
Supplied with cap and O-Ring.
Ordering information: For replacement caps, see 89075-248; for O-Rings, see 89060-994.
Customers Who Bought This Also Bought