SP Bel-Art Dialysis Bag Clip Holders, Bel-Art Products, a part of SP
Catalog #: 11215-308
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Dialysis Bag Clip Holders
- Cat. no.:11215-308
- Supplier No.:182370000
Specifications
About this item
Suspend dialysis bags in large beakers or pitchers up to 0.63 cm (1/4") thick with these strong clips.
- Smooth polypropylene construction
- Autoclavable
- Easy to mark with sequence numbers
Bags slide easily into the slot and then the holder can be clipped onto the container’s edge. Smooth polypropylene surface may be marked with identifying sequence numbers.
Customers Who Bought This Also Bought