SP Bel-Art Cup-Type Stoppers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
These polyethylene cup-type stoppers fit tightly yet remove easily.
- Polyethylene construction
- Tight fit, yet easy to remove
- Cup style allows them to double as micro containers
They also double as containers for micro work. All are 26 mm high.
Customers Who Bought This Also Bought