n-Hexane, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MERCK, S.A. DE C.V. SV
Certificates
About this item
Synonyms: Hexyl hydride,Hexanen
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MW:86.18 g/mol
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–22 °C
MDL Number:MFCD00009520
ADR:3,II
Merck Index:13,04712
:n-Hexane
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ300
:Boiling Pt:
:Boiling Pt: 69 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –94.3 °C
:Flash Pt:
:Flash Pt: –22 °C
:M.W. 86.18 g/mol
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
:3
:II
:–22 °C
:GHS02
:GHS08
:GHS07
:GHS09
:MQ300
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Purity (GC):≥ 96.0 %
- Purity Σ hexane isomers + methylcyclopentane (GC):≥ 98.5 %
- Identity (IR):conforms
- Color:≤ 10 Hazen
- Water-soluble titrable acid:≤ 0.0003 meq/g
- Refractive index (n 20/D):1.375 - 1.376
- Density (d 20 °C/20 °C):0.659 - 0.663
- Boiling range (min.95 %):67 - 69 °C
- Transmission (between 260 nm and 420 nm):≥ 97 %
- Thiophene:conforms
- Aromatics (as benzene):≤ 0.01 %
- Sulfur compounds (as S):≤ 0.005 %
- Readily carbonizable substances:conforms
- Al (Aluminium):≤ 0.00005 %
- B (Boron):≤ 0.000002 %
- Ba (Barium):≤ 0.00001 %
- Ca (Calcium):≤ 0.00005 %
- Cd (Cadmium):≤ 0.000005 %
- Co (Cobalt):≤ 0.000002 %
- Cr (Chromium):≤ 0.000002 %
- Cu (Copper):≤ 0.000002 %
- Fe (Iron):≤ 0.00001 %
- Mg (Magnesium):≤ 0.00001 %
- Mn (Manganese):≤ 0.000002 %
- Ni (Nickel):≤ 0.000002 %
- Pb (Lead):≤ 0.00001 %
- Sn (Tin):≤ 0.00001 %
- Zn (Zinc):≤ 0.00001 %
- Evaporation residue:≤ 0.001 %
- Water:≤ 0.01 %
- Supelco®, n-Hexane, EMSURE® ACS, Reag. Ph. Eur. for analysis:
Specification Test Results
Related Information
Certificates
Search by Lot NumberMilliporeSigma EasyGrip Full Control Ergonomic HDPE Bottlesfor Acids, Bases and Solvents
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought