Formic acid 98-100%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Methanoic acid
Formula:HCO₂H
MW:46.03 g/mol
MDL Number:MFCD00003297
CAS Number:64-18-6
:46.03
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98-100
:M-Clarity™ quality level = MQ300
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 8.2 to 8.4 °C
:Flash Pt:
:Flash Pt:
:Density: 1,22 g/cm³ (25 °C)
:M.W. 46.03 g/mol
:Formic acid
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
:8
:II
:1.22
:48
:GHS02
:GHS06
:GHS05
:MQ300
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (alkalimetric):≥ 98.0 %
- Color:≤ 10 Hazen
- Acetic acid (CH₃COOH):≤ 500 ppm
- Chloride (Cl):≤ 5 ppm
- Sulfate (SO₄):≤ 5 ppm
- Sulfite (SO₃):≤ 10 ppm
- Heavy metals (as Pb):≤ 10 ppm
- Ag (Silver):≤ 0.020 ppm
- Al (Aluminium):≤ 0.050 ppm
- Ba (Barium):≤ 0.050 ppm
- Be (Beryllium):≤ 0.020 ppm
- Bi (Bismuth):≤ 0.10 ppm
- Ca (Calcium):≤ 0.20 ppm
- Cd (Cadmium):≤ 0.050 ppm
- Co (Cobalt):≤ 0.020 ppm
- Cr (Chromium):≤ 0.050 ppm
- Cu (Copper):≤ 0.020 ppm
- Fe (Iron):≤ 2.0 ppm
- Ge (Germanium):≤ 0.050 ppm
- K (Potassium):≤ 0.10 ppm
- Li (Lithium):≤ 0.020 ppm
- Mg (Magnesium):≤ 0.50 ppm
- Mn (Manganese):≤ 0.050 ppm
- Mo (Molybdenum):≤ 0.020 ppm
- NH₄ (Ammonium):≤ 10 ppm
- Na (Sodium):≤ 0.50 ppm
- Ni (Nickel):≤ 0.050 ppm
- Pb (Lead):≤ 0.020 ppm
- Sr (Strontium):≤ 0.020 ppm
- Ti (Titanium):≤ 0.10 ppm
- Tl (Thallium):≤ 0.050 ppm
- V (Vanadium):≤ 0.050 ppm
- Zn (Zinc):≤ 0.050 ppm
- Zr (Zirconium):≤ 0.10 ppm
- Evaporation residue:≤ 10 ppm
- Dilution test:passes test
- Supelco®, Formic acid 98-100%, EMSURE® ACS, Reag. Ph. Eur. for analysis:
Specification Test Results
Related Information
Certificates
Search by Lot NumberEMD Millipore Solutions for Sample Digestion Flyer EMD Polar Compounds Merck Inorganic on tap, Withdrawal Systems and Safety Accessories for Acids and Bases MilliporeSigma Acid in Safebreak Bottles.pdf
Product Literature
ghs_1.00264
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought