Acetic acid ≥96%, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Vinegar acid,Methanecarboxylic acid
Formula:H₃CCOOH
MW:60.05 g/mol
MDL Number:MFCD00036152
CAS Number:64-19-7
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:96
:M-Clarity™ quality level = MQ300
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt:
:M.W. 60.05 g/mol
EINECS:200-580-7
:17
:118
UN:2789
:8
:II
:38.5
:GHS02
:GHS05
Storage Temperature:Ambient
:MQ300
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (alkalimetric):≥ 96.0 %
- Color:≤ 10 Hazen
- Acetaldehyde:≤ 2 ppm
- Chloride (Cl):≤ 0.5 ppm
- Phosphate (PO₄):≤ 0.5 ppm
- Sulfate (SO₄):≤ 0.5 ppm
- Ag (Silver):≤ 0.010 ppm
- Al (Aluminium):≤ 0.050 ppm
- As (Arsenic):≤ 0.01 ppm
- Au (Gold):≤ 0.010 ppm
- B (Boron):≤ 0.100 ppm
- Ba (Barium):≤ 0.010 ppm
- Be (Beryllium):≤ 0.020 ppm
- Bi (Bismuth):≤ 0.100 ppm
- Ca (Calcium):≤ 0.200 ppm
- Cd (Cadmium):≤ 0.050 ppm
- Co (Cobalt):≤ 0.010 ppm
- Cr (Chromium):≤ 0.020 ppm
- Cu (Copper):≤ 0.020 ppm
- Fe (Iron):≤ 0.100 ppm
- Ga (Gallium):≤ 0.050 ppm
- Ge (Germanium):≤ 0.050 ppm
- In (Indium):≤ 0.050 ppm
- K (Potassium):≤ 0.100 ppm
- Li (Lithium):≤ 0.010 ppm
- Mg (Magnesium):≤ 0.050 ppm
- Mn (Manganese):≤ 0.010 ppm
- Mo (Molybdenum):≤ 0.020 ppm
- Na (Sodium):≤ 0.500 ppm
- Ni (Nickel):≤ 0.020 ppm
- Pb (Lead):≤ 0.020 ppm
- Pt (Platinum):≤ 0.100 ppm
- Sn (Tin):≤ 0.050 ppm
- Sr (Strontium):≤ 0.010 ppm
- Ti (Titanium):≤ 0.100 ppm
- Tl (Thallium):≤ 0.050 ppm
- V (Vanadium):≤ 0.010 ppm
- Zn (Zinc):≤ 0.050 ppm
- Zr (Zirconium):≤ 0.100 ppm
- Indifference against chromic acid:conforms
- Substances reducing potassium permanganate:conforms
- Evaporation residue:≤ 5 ppm
- Dilution test:conforms
- Date of expiry:see product label
- Supelco®, Acetic acid ≥96%, EMSURE® for analysis:
Specification Test Results
Related Information
Certificates
Search by Lot NumberEMD Millipore Bacteriology Tissue Diagnostics EMD Millipore Solutions for Sample Digestion Flyer Merck Inorganic on tap, Withdrawal Systems and Safety Accessories for Acids and Bases MilliporeSigma EasyGrip Full Control Ergonomic HDPE Bottlesfor Acids, Bases and Solvents
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought