Buchner Funnels, Globe Scientific
Catalog #: 10027-864
Supplier: Globe Scientific
Specifications
- Cat. no.:10027-864
- Top Diameter:90 mm
- Supplier No.:600441
Specifications
About this item
Molded from Polypropylene (PP), these funnels are unbreakable and resistant to corrosion and heat
- Polypropylene (PP)
- Autoclavable
- Food safe
These funnels are approved for use with food products.
Two piece construction allows top and bottom to be separated for easy cleaning. The funnels are fully autoclavable and feature excellent chemical resistance.
Customers Who Bought This Also Bought