Preparative TLC Plates, Agela Technologies
Supplier: Avantor
Certificates
About this item
These thin layer chromatography plates are used for a variety of chemistries
They are flexible and glass-backed for easy use. When matching with Claricep flash cartridges, the TLC plates serve as a tool for reaction monitoring and method development based on Rf values for flash chromatography.
Customers Who Bought This Also Bought