Alcohol, anhydrous, Reagent Grade for histology, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Reagent alcohol
MDL Number:MFCD00003568
CAS Number:64-17-5
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:Alcohol
EINECS:200-578-6
UN:1170
:3
:II
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Color (APHA):≤10
- Specific Gravity at 25°C:0.785 - 0.795
- Residue after Evaporation:≤0.0100 %
- Water (H₂O):≤0.5 %
- Assay Specially Denatured Alcohol Formula 3A (200 proof)(v/v):94.0 - 96.0 %
- Isopropyl Alcohol (2-propanol)(v/v):4.0 - 6.0 %
- SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200proof ethanol.:
- For General Purpose Reagent Use Only.:
- NOTE: Intended for nonfuel end purposes only.:
- J.T.Baker®, Alcohol, anhydrous, Reagent Grade for histology:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
sds_BAKRA478
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought