BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
Supplier: BD
Certificates
About this item
This dehydrated culture media is used for isolating and differentiating lactose-fermenting from lactose-nonfermenting gram-negative enteric bacilli.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.