ContactUsLinkComponent
KOD Xtreme™ Hot Start DNA Polymerase, MilliporeSigma
KOD Xtreme™ Hot Start DNA Polymerase, MilliporeSigma
Catalog #: CA95042-700
Supplier:  MilliporeSigma
CAS Number:  
KOD Xtreme™ Hot Start DNA Polymerase, MilliporeSigma
Catalog #: CA95042-700
Supplier:  MilliporeSigma
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    KOD Xtreme™ Hot Start DNA Polymerase, 2X Xtreme Buffer
  • Size:
    200 Units
  • Cat. no.:
    CA95042-700
  • Supplier No.:
    71975-3
Specifications
About this item
The KOD Xtreme™ Hot Start DNA Polymerase is a ready-to-use reaction or routine PCR amplification, and optimized for difficult targets
Efficiently amplifies up to 90% GC-content DNA templates, genomic targets up to 24 kbp and phage/Lambda targets up to 40 kbp.
Customers Who Bought This Also Bought