To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:karyopherin alpha 1 (importin alpha 5)
- Clonality:Polyclonal
- Gene ID:P52294
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:KPNA1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:KPNA1
- Storage buffer:KPNA1 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:KPNA1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:KPNA1 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human KPNA1. The immunogen is located within amino acids 30 - 80 of KPNA1.
- Tested applications:Immunocytochemistry
- Purification:KPNA1 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89417-608
- Supplier No.:5981
Specifications
About this item
KPNA1 Antibody: Karyopherin, a cytosolic and heterodimeric protein complex consisting of alpha and beta subunits, is responsible for targeting proteins with nuclear localization signals to the nuclear pore complex by an energy requiring, Ran-dependent mechanism. The alpha subunit and imported substrate enter the nucleus and accumulate in the nucleoplasm, while the beta subunit accumulates at the NPC. KPNA1, also known as importin alpha 5, is the alpha subunit of karyopherin, which forms a complex with importin subunit beta-1 and functions in nuclear protein import as an adapter protein for nuclear receptor KPNB1. It is ubiquitously expressed and polyubiquitinated in the presence of RAG1. KPNA1 interacts with various virus nucleoproteins, including those of Ebola and influenza.
Type: Primary
Antigen: KPNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat