To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:receptor accessory protein 2
- Clonality:Polyclonal
- Gene ID:Q9BRK0
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:REEP2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:REEP2
- Storage buffer:REEP2 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:REEP2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:REEP2 antibody was raised against a 15 amino acid synthetic peptide near the center of human REEP2. The immunogen is located within amino acids 150 - 200 of REEP2.
- Tested applications:E, WB, IHC, IF
- Purification:REEP2 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89417-298
- Supplier No.:5571
Specifications
About this item
REEP2 Antibody: Mammalian odorant receptors require accessory proteins such as RTP1 and RTP2 for functional cell surface expression. Receptor expression-enhancing protein (REEP) family members are transmembrane proteins which interact with odorant receptor proteins and may enhance the odorant receptor responses to odorants. Recently studies have shown other chemosensory receptors such as bitter taste receptors are also influenced by RTP and REEP family members.
Type: Primary
Antigen: REEP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat