To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:transmembrane protein 18
- Clonality:Polyclonal
- Gene ID:Q96B42
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TMEM18
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TMEM18
- Storage buffer:TMEM18 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:TMEM18 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:TMEM18 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human TMEM18. The immunogen is located within the last 50 amino acids of TMEM18.
- Tested applications:E, WB, IHC, IF
- Purification:TMEM18 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89416-904
- Supplier No.:5043
Specifications
About this item
TMEM18 Antibody: Recent studies have examined how overexpression of certain genes enhances the tropism of neural stem cells (NSCs) for gliomas. Overexpression of Transmembrane protein 18 (TMEM18) provides NSCs and neural precursors an increased migration capacity towards glioblastoma cells in vitro and in the rat brain, while functional inactivation of the gene resulted in an almost complete loss of migration activity, suggesting that TMEM18 could be a potential target in NSC-based glioma therapy. Other recent results suggest that among others, the TMEM18 loci is associated with higher than normal body mass index (BMI) and obesity, indicating there may be a CNS-related role in predisposition to obesity. Multiple isoforms of TMEM18 are known to exist.
Type: Primary
Antigen: TMEM18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat