To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:dispatched homolog 2 (Drosophila)
- Clonality:Polyclonal
- Gene ID:A7MBM2
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DISP2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:DISP2
- Storage buffer:Disp2 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:Disp2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:Disp2 antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human Disp2. The immunogen is located within amino acids 1210 - 1260 of Disp2.
- Tested applications:E, WB
- Purification:Disp2 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89416-790
- Supplier No.:4883
Specifications
About this item
Disp2 Antibody: Disp2 is the second of three known homologs of the D. melanogaster protein Dispatched. It is a multi-transmembrane protein containing two PTCH/DISP domains and is thought to be involved in the release of lipid-anchored Hedgehog from producing cells. Hedgehog is a major player in signaling pathways during embryogenesis, tissue regeneration, and carcinogenesis and the Disp proteins have been implicated in these pathways. Recently, it has been shown that Disp2 is translationally regulated by the microRNA miR-214 in zebrafish. Expression of this miRNA decreased Disp2 promoter activity in vitro and its overexpression in zebrafish resulted in a phenotype identical to that observed by Disp2 mutants. At least two isoforms of Disp2 are known to exist. This Disp2 antibody is predicted to not cross-react with Disp1 or Disp3.
Type: Primary
Antigen: DISP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat