To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:amyloid beta (A4) precursor protein binding, family A, member 1
- Clonality:Polyclonal
- Gene ID:B2RUJ5
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:Apba1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Apba1
- Storage buffer:UROP11 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:UROP11 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:UROP11 antibody was raised against a 12 amino acid synthetic peptide near the center of the mouse UROP11. The immunogen is located within the first 50 amino acids of UROP11.
- Tested applications:E, WB
- Purification:UROP11 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89416-592
- Supplier No.:4615
Specifications
About this item
UROP11 Antibody: Upregulated obese product 11 (UROP11) was isolated from subtractive library for mouse hypothalamus. It is detected in numerous tissues and is upregulated in the hypothalamus and adipocytes in obese mice. UROP11 is also observed in human neural and lymphoid tissues; its expression appears to be regulated by the level of lymphoid activation. Urop11 expression is strongly upregulated both in vivo in mouse hypothalamus and in vitro in mouse neural cell lines after treatment with leptin, indicating Urop11 is a target of leptin activity and thus may play a role in obesity.
Type: Primary
Antigen: Apba1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat