To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:fasciculation and elongation protein zeta 2 (zygin II)
- Clonality:Polyclonal
- Gene ID:Q9UHY8
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:FEZ2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:FEZ2
- Storage buffer:FEZ2 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:FEZ2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:FEZ2 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human FEZ2. The immunogen is located within the last 50 amino acids of FEZ2.
- Tested applications:E, WB, IF
- Purification:FEZ2 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89416-432
- Supplier No.:4403
Specifications
About this item
FEZ2 Antibody: Fasciculation and elongation protein zeta-2 (FEZ2) is a homolog to the mammalian FEZ1, itself an ortholog of the C. elegans UNC-76. In contrast to FEZ1, FEZ2 mRNA is widely expressed in mouse tissues. FEZ2 interacts with protein kinase C (PKC)-zeta substrate and induces neurite extension of PC12 cells when co-expressed with a constitutively active form of PKC-zeta, suggesting FEZ2 may play an important role in the morphological changes of various cells by associating with PKC-zeta in a tissue non-specific manner. FEZ2 can interact with FEZ1 through its c-terminal regions and especially its coiled-coil region. At least two isoforms of FEZ2 are known to exist.
Type: Primary
Antigen: FEZ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat