To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:programmed cell death 1 ligand 2
- Antigen symbol:PDCD1LG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9BQ51
- Antigen synonyms:PDCD1LG2
- Storage buffer:PD-L2 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 30 kDa Observed: 32 kDa
- Storage temperature:PD-L2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:PD-L2 antibody was raised against a 16 amino acid synthetic peptide from near the center of human PD-L2. The immunogen is located within amino acids 140 - 190 of PD-L2.
- Tested applications:E, WB, IHC, IF
- Purification:PD-L2 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89416-220
- Supplier No.:4063
PDL-2 Antibody: Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by co gnate peptides bound to MHC molecules on antigen-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2, both of which are thought act as a negative regulator of T cell activation. However, it has been suggested that PDL-2 can act to stimulate an immunogenic response through and alternative receptor from PD-1. At least three isoforms of PDL-2 are known to exist; this antibody is specific to the largest isoform. PDL-2 antibody has no cross-reactivity to PDL-2.
Type: Primary
Antigen: PDCD1LG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat