To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:RPA interacting protein
- Clonality:Polyclonal
- Gene ID:Q86UA6
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:RPAIN
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:RPAIN
- Storage buffer:RPA Interacting Protein Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:RPA Interacting Protein antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:RPA Interacting Protein antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human RPA Interacting Protein. The immunogen is located within the first 50 amino acids of RPA Interacting Protein.
- Tested applications:E, WB
- Purification:RPA Interacting Protein Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89416-178
- Supplier No.:4015
Specifications
About this item
RPA Interacting Protein Antibody: Replication protein A (RPA) is a single-stranded-DNA binding protein involved in numerous eukaryotic DNA processes including replication, repair and recombination. RPA interacting protein (RPA IP) has been identified as an adapter protein that is involved in RPA nuclear import instead of the prototypical importin proteins that normally mediate nuclear import. Multiple isoforms of RPA IP are known to exist, with the longest isoform localized to the cytoplasm. Isoform 2 is sumoylated and is located in the PML nuclear body within the nucleus. It has been suggested that this isoform mediates the localization of the RPA complex into the PML nuclear body, thereby participating in RPA function in DNA metabolism.
Type: Primary
Antigen: RPAIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat