To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fas apoptotic inhibitory molecule 2
- Clonality:Polyclonal
- Gene ID:Q9BWQ8
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:FAIM2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:FAIM2
- Storage buffer:FAIM2 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:FAIM2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:FAIM2 antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human FAIM2. The immunogen is located within the first 50 amino acids of FAIM2.
- Tested applications:E, WB, IHC
- Purification:FAIM2 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:89415-418
- Supplier No.:2285
Specifications
About this item
LFG Antibody: Programmed cell death regulates a number of biological processes such as normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. LFG is a recently identified protein that can inhibit the apoptotic signal transduced by the Fas receptor but not from the related tumor necrosis factor-alpha death signal. In this respect, LFG is functionally similar to the anti-apoptotic proteins FAIM, FLIP and Bcl-xL. LFG, a seven membrane spanning protein, can bind the Fas receptor but does not regulate Fas expression or inhibit binding of FADD to Fas. LFG is widely distributed, but highly expressed in the hippocampus and other neural tissues. LFG was also identified as the neural membrane protein 35 (NMP35) and its expression is known to be regulated by the Phosphatidylinositol 3-kinase-Akt/PKB pathway.
Type: Primary
Antigen: FAIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat