Order Entry
ContactUsLinkComponent
Anti-MCL1 Rabbit Polyclonal Antibody
Anti-MCL1 Rabbit Polyclonal Antibody
Catalog #: 89415-810
Supplier:  Prosci
MCL-1 0.1MG
Anti-MCL1 Rabbit Polyclonal Antibody
Catalog #: 89415-810
Supplier:  Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    myeloid cell leukemia sequence 1 (BCL2-related)
  • Clonality:
    Polyclonal
  • Gene ID:
    Q07820
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    MCL1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    0.1 mg
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    MCL1
  • Storage buffer:
    Mcl-1 Antibody is supplied in PBS containing 0.02% sodium azide.
  • Storage temperature:
    Mcl-1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    4 °C
  • Immunogen:
    Mcl-1 antibody was raised against a peptide corresponding to 16 amino acids near the center of human Mcl-1. The immunogen is located within amino acids 110 - 160 of Mcl-1.
  • Tested applications:
    Immunocytochemistry
  • Purification:
    Mcl-1 Antibody is Ion exchange chromatography purified.
  • Cat. no.:
    89415-810
  • Supplier No.:
    3481

Specifications

About this item

Mcl-1 Antibody: Myeloid cell leukimia-1 (Mcl-1) is a member of the Bcl-2 family of proteins that can act to promote cell survival. While the mechanism by which Mcl-1 inhibits apoptosis is not known, it is thought that it may heterodimerize and neutralize pro-apoptotic members of the Bcl-2 family such as Bim or Bak. Mcl-1 was originally identified in differentiating myeloid cells, but has since been shown to be expressed in multiple cell types. Mcl-1 is essential for embryogenesis and for the development and maintenance of B and T lymphocytes in animals. Mcl-1 exists as at least two distinct isoforms designated Mcl-1L and Mcl-1S. In marked contrast to the larger isoform of Mcl-1, overexpression of Mcl-1S promotes cell death.

Type: Primary
Antigen: MCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought